accessibility menu, dialog, popup

UserName

NH2H2N

CAS RN 1761-71-3

IUPAC Name: 4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
Synonyms: 4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
Molecular Weight (g/mol): 210.37
Molecular Formula: C13H26N2
InChi Key: DZIHTWJGPDVSGE-UHFFFAOYSA-N
SMILES: NC1CCC(CC2CCC(N)CC2)CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)