missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(cyclohexylamine), 95%

Product Code. 15244654 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15244654 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15244654 Supplier Thermo Scientific Acros Supplier No. 159342500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1761-71-3 | C13H26N2 | 210.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1761-71-3
Melting Point 36.5°C
Boiling Point 325.0°C to 327.0°C
Flash Point 159°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C13H26N2
Linear Formula CH2(C6H10NH2)2
Quantity 250 g
Synonym 4,4'-methylenedicyclohexanamine, 4,4'-diaminodicyclohexylmethane, 4,4'-methylenebis cyclohexylamine, cyclohexanamine, 4,4'-methylenebis, wandamin hm, bis 4-aminocyclohexyl methane, bis p-aminocyclohexyl methane, 4,4'-diaminodicyclohexyl methane, pacm 20, cyclohexylamine, 4,4'-methylenebis
Solubility Information Solubility in water: 5g/L (20°C)
InChI Key DZIHTWJGPDVSGE-UHFFFAOYSA-N
SMILES C1CC(CCC1CC2CCC(CC2)N)N
IUPAC Name 4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
Molecular Weight (g/mol) 210.36
PubChem CID 15660
Formula Weight 210.36
Percent Purity 95%
Chemical Name or Material 4, 4'-Methylenebis(cyclohexylamine)
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4'-Methylenebis(cyclohexylamine)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.