accessibility menu, dialog, popup

UserName

CAS RN 17635-26-6

H3CNNH3COO

CAS RN 17635-26-6

IUPAC Name: 2,3-dimethylquinoxaline-6-carboxylate
Synonyms: 2,3-dimethylquinoxaline-6-carboxylate
Molecular Weight (g/mol): 201.21
Molecular Formula: C11H9N2O2
InChi Key: RCACNAWRFYUKLC-UHFFFAOYSA-M
SMILES: CC1=C(C)N=C2C=C(C=CC2=N1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results