CAS RN 176721-02-1
CAS RN 176721-02-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2,3-Dimethylphenyl)[1-(trityl)-1H-imidazol-4-yl]methanone, TRC
CAS: 176721-02-1 Molecular Formula: C31H26N2O Molecular Weight (g/mol): 442.55 Synonym: (2,3-Dimethylphenyl)[1-(triphenylmethyl)-1H-imidazol-4-yl]methanone SMILES: Cc1cccc(C(=O)c2cn(cn2)C(c3ccccc3)(c4ccccc4)c5ccccc5)c1C