accessibility menu, dialog, popup

UserName

BrH3COOP

CAS RN 1779-58-4

IUPAC Name: (2-methoxy-2-oxoethyl)triphenylphosphanium bromide
Synonyms: (2-methoxy-2-oxoethyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 415.27
Molecular Formula: C21H20BrO2P
InChi Key: VCWBQLMDSMSVRL-UHFFFAOYSA-M
SMILES: [Br-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results