accessibility menu, dialog, popup

UserName

NH2NS

CAS RN 1779-81-3

IUPAC Name: 4,5-dihydro-1,3-thiazol-2-amine
Synonyms: 2-aminothiazoline 氨噻唑啉 usaf PD-57
Molecular Weight (g/mol): 102.16
Molecular Formula: C3H6N2S
InChi Key: REGFWZVTTFGQOJ-UHFFFAOYSA-N
SMILES: NC1=NCCS1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)