missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-thiazoline, 97%

Product Code. 11936781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11936781 100 g Glass bottle 100g
11926781 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11936781 Supplier Thermo Scientific Acros Supplier No. 438851000
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1779-81-3 | C3H6N2S | 102.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1779-81-3
Molecular Formula C3H6N2S
Molecular Weight (g/mol) 102.15
InChI Key REGFWZVTTFGQOJ-UHFFFAOYSA-N
Synonym 2-aminothiazoline, 2-amino-2-thiazoline, 2-iminothiazolidine, 4,5-dihydrothiazol-2-amine, 2-thiazolidinimine, 2-amino-4,5-dihydrothiazole, 2-thiazolamine, 4,5-dihydro, 2-thiazoline, 2-amino, usaf pd-57, 2-amino-4,5-dihydrothiazoline
PubChem CID 15689
ChEBI CHEBI:40889
IUPAC Name 4,5-dihydro-1,3-thiazol-2-amine
SMILES C1CSC(=N1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 77°C to 81°C
Color White
Boiling Point 221.8°C
Flash Point 87.9°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: soluble
Formula Weight 102.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2-thiazoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-2-thiazoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.