accessibility menu, dialog, popup

UserName

CH3OOFF

CAS RN 178312-47-5

IUPAC Name: ethyl 4,4-difluorocyclohexane-1-carboxylate
Synonyms: ethyl 4,4-difluorocyclohexane-1-carboxylate
Molecular Weight (g/mol): 192.21
Molecular Formula: C9H14F2O2
InChi Key: HZZDWLBBNSDYQM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCC(F)(F)CC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results