missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4-difluorocyclohexanecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 11389969
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389969 5 g 5g
11369969 250 mg 250mg
11379969 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389969 Supplier Thermo Scientific Alfa Aesar Supplier No. H25754.06
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 178312-47-5 | Molecular Formula: C9H14F2O2 | Molecular Weight: 192.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178312-47-5
Molecular Formula C9H14F2O2
Molecular Weight (g/mol) 192.206
MDL Number MFCD03788494
InChI Key HZZDWLBBNSDYQM-UHFFFAOYSA-N
PubChem CID 2779138
IUPAC Name ethyl 4,4-difluorocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCC(CC1)(F)F

Specifications

Boiling Point 84°C to 86°C (8 mmHg)
Quantity 5 g
Synonym ethyl 4,4-difluorocyclohexanecarboxylate, cyclohexanecarboxylic acid, 4,4-difluoro-, ethyl ester, ethyl4,4-difluorocyclohexanecarboxylate, 4,4-difluoro-1-ethoxycarbonyl cyclohexane, 4,4-difluoro-cyclohexanecarboxylic acid ethyl ester, ethyl 4,4-difluorocyclohexane carboxylate, ethyl-4,4-difluorocyclohexane carboxylate, cyclohexanecarboxylic acid, 4,4-difluoro-, ethyl ester 9ci, acmc-209xsu, 4-carbethoxy-1,1-difluorocyclohexane
Formula Weight 192.21
Percent Purity 97%
Chemical Name or Material Ethyl 4,4-difluorocyclohexanecarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.