CAS RN 1789818-21-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,5-Dimethyl-N-2-propen-1-yl-3-thiophenecarboxamide, TRC
CAS: 1789818-21-8 Molecular Formula: C10 H13 N O S Molecular Weight (g/mol): 195.281 Synonym: Des-TMS silthiofam,Silthiofam metabolite IUPAC Name: 4,5-dimethyl-N-prop-2-enylthiophene-3-carboxamide SMILES: Cc1scc(C(=O)NCC=C)c1C