missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-N-2-propen-1-yl-3-thiophenecarboxamide, TRC

Product Code. 30133436
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133436 500 mg 500mg
30011438 250 mg 250mg
30011034 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133436 Supplier Toronto Research Chemicals Supplier No. TRCD222555500MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1789818-21-8
Molecular Formula C10 H13 N O S
Molecular Weight (g/mol) 195.281
Synonym Des-TMS silthiofam, Silthiofam metabolite
IUPAC Name 4,5-dimethyl-N-prop-2-enylthiophene-3-carboxamide
SMILES Cc1scc(C(=O)NCC=C)c1C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H13NOS/c1-4-5-11-10(12)9-6-13-8(3)7(9)2/h4,6H,1,5H2,2-3H3,(H,11,12)
Formula Weight 195.0718
Chemical Name or Material Silthiofam-des(trimethylsilyl)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.