accessibility menu, dialog, popup

UserName

H3COOHCl

CAS RN 18113-03-6

IUPAC Name: 2-chloro-4-methoxyphenol
Synonyms: 2-chloro-4-methoxyphenol
Molecular Weight (g/mol): 158.58
Molecular Formula: C7H7ClO2
InChi Key: GNVRRKLFFYSLGT-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C(Cl)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)