accessibility menu, dialog, popup

UserName

CAS RN 181418-64-4

IUPAC Name: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) titanium
Synonyms: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) titanium
Molecular Weight (g/mol): 600.7
Molecular Formula: C33H60O6Ti
InChi Key: GDCBFXOQZJSCHZ-UHFFFAOYSA-N
SMILES: [Ti].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)