missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)titanium(III), Thermo Scientific™

Product Code. 11338578
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338578 25 g 25g
11328578 1 g 1g
11318578 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338578 Supplier Thermo Scientific Alfa Aesar Supplier No. 040621.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181418-64-4
Molecular Formula C33H60O6Ti
Molecular Weight (g/mol) 600.704
MDL Number MFCD00145466
InChI Key GDCBFXOQZJSCHZ-LWTKGLMZSA-N
Synonym tris 2,2,6,6-tetramethyl-3,5-heptanedionato titanium iii
PubChem CID 118856175
IUPAC Name (Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;titanium
SMILES CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[Ti]

Specifications

Color Black to Purple
Boiling Point 75°C (0.1 mmHg)(sublimation)
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 597.7
Physical Form Crystalline
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)titanium(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.