accessibility menu, dialog, popup

UserName

CAS RN 18179-39-0

CH3OOIOH

CAS RN 18179-39-0

IUPAC Name: methyl 2-hydroxy-4-iodobenzoate
Synonyms: methyl 2-hydroxy-4-iodobenzoate
Molecular Weight (g/mol): 278.05
Molecular Formula: C8H7IO3
InChi Key: WUFUURSWOJROKY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results