accessibility menu, dialog, popup

UserName

ClONN

CAS RN 1822-94-2

IUPAC Name: 5-(chloromethyl)-3-phenyl-1,2,4-oxadiazole
Synonyms: 5-(chloromethyl)-3-phenyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 194.62
Molecular Formula: C9H7ClN2O
InChi Key: XMSYUUPMPGDIOE-UHFFFAOYSA-N
SMILES: ClCC1=NC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results