missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-3-phenyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Product Code. 10216422
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10216422 1 g Amber Glass Bottle 1g
10152844 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216422 Supplier Thermo Scientific Maybridge Supplier No. RF01832DA
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1822-94-2
Molecular Formula C9H7ClN2O
Molecular Weight (g/mol) 194.618
InChI Key XMSYUUPMPGDIOE-UHFFFAOYSA-N
PubChem CID 966714
IUPAC Name 5-(chloromethyl)-3-phenyl-1,2,4-oxadiazole
SMILES C1=CC=C(C=C1)C2=NOC(=N2)CCl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-chloromethyl-3-phenyl-1,2,4-oxadiazole, 5-chloromethyl-3-phenyl-1,2,4 oxadiazole, 3-phenyl-5-chloromethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-chloromethyl-3-phenyl, pubchem16349, aronis24545, 5-chloromethyl-3-phenyl-1,2,4-oxadiazol, 1,2,4-oxadiazole,5-chloromethyl-3-phenyl
Formula Weight 194.62
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)-3-phenyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.