CAS RN 1829-37-4
IUPAC Name:
2-bromophenyl acetate
Synonyms:
2-bromophenyl acetate
Molecular Weight (g/mol):
215.05
Molecular Formula:
C8H7BrO2
InChi Key:
BEHBHYYPTOHUHX-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=CC=CC=C1Br
1
–
1
of
1
results
1
2-Bromophenyl acetate, 98%, Thermo Scientific™
CAS: 1829-37-4 Molecular Formula: C8H7BrO2 Molecular Weight (g/mol): 215.05 InChI Key: BEHBHYYPTOHUHX-UHFFFAOYSA-N Synonym: o-bromophenyl acetate,2-bromophenol acetate,1-acetoxy-2-bromobenzene,2-bromophenyl acetate,o-acetoxybromobenzene,phenol, 2-bromo-, acetate,phenol, o-bromo-, acetate,pubchem3055,2-bromo-phenol acetate PubChem CID: 74586 IUPAC Name: (2-bromophenyl) acetate SMILES: CC(=O)OC1=CC=CC=C1Br