CAS RN 184970-29-4
IUPAC Name:
[3-fluoro-5-(trifluoromethyl)phenyl]methanol
Synonyms:
[3-fluoro-5-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol):
194.13
Molecular Formula:
C8H6F4O
InChi Key:
LTWAIZVPDMHGOE-UHFFFAOYSA-N
SMILES:
OCC1=CC(F)=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Fluoro-5-(trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™
CAS: 184970-29-4 Molecular Formula: C8H6F4O Molecular Weight (g/mol): 194.129 MDL Number: MFCD00061169 InChI Key: LTWAIZVPDMHGOE-UHFFFAOYSA-N Synonym: 3-fluoro-5-trifluoromethyl benzyl alcohol,3-fluoro-5-trifluoromethyl benzylalcohol,3-fluoro-5-trifluoromethyl phenyl methanol,benzenemethanol, 3-fluoro-5-trifluoromethyl,5-fluoro-3-trifluoromethyl phenyl methan-1-ol,pubchem4633,acmc-209xc7,ksc496c0f PubChem CID: 589176 IUPAC Name: [3-fluoro-5-(trifluoromethyl)phenyl]methanol SMILES: C1=C(C=C(C=C1C(F)(F)F)F)CO