accessibility menu, dialog, popup

UserName

CAS RN 186405-37-8

FFFSOON

CAS RN 186405-37-8

IUPAC Name: 2-[4-(trifluoromethyl)benzenesulfonyl]acetonitrile
Synonyms: 2-[4-(trifluoromethyl)benzenesulfonyl]acetonitrile
Molecular Weight (g/mol): 249.21
Molecular Formula: C9H6F3NO2S
InChi Key: AGVHYIFQWMKQLX-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)S(=O)(=O)CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results