accessibility menu, dialog, popup

UserName

CAS RN 186517-00-0

IUPAC Name: 3-chloro-2-fluoro-6-(trifluoromethyl)benzonitrile
Synonyms: 3-chloro-2-fluoro-6-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 223.55
Molecular Formula: C8H2ClF4N
InChi Key: AVIILPWTQMSBOW-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC(=C1C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results