accessibility menu, dialog, popup

UserName

OOHI

CAS RN 18698-96-9

IUPAC Name: 2-(2-iodophenyl)acetic acid
Synonyms: benzeneacetic acid, 2-iodo-
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: IUHXGZHKSYYDIL-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1I
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)