accessibility menu, dialog, popup

UserName

CH3NNOO

CAS RN 18699-87-1

IUPAC Name: 2-methyl-3-nitropyridine
Synonyms: 2-methyl-3-nitropyridine
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: CCFGTKQIRWHYTB-UHFFFAOYSA-N
SMILES: CC1=NC=CC=C1[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results