accessibility menu, dialog, popup

UserName

H3COO(E)

CAS RN 1871-67-6

IUPAC Name: (2E)-oct-2-enoate
Synonyms: (2E)-oct-2-enoate
Molecular Weight (g/mol): 141.19
Molecular Formula: C8H13O2
InChi Key: CWMPPVPFLSZGCY-VOTSOKGWSA-M
SMILES: CCCCC\C=C\C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results