missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Octenoic Acid, TRC

Product Code. 30453719
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453719 100 mg 100mg
30450818 1 g -
30452675 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453719 Supplier Toronto Research Chemicals Supplier No. TRCT705480100MG
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-67-6
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.2
IUPAC Name (E)-oct-2-enoic acid
SMILES CCCCC\C=C\C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H14O2/c1-2-3-4-5-6-7-8(9)10/h6-7H,2-5H2,1H3,(H,9,10)/b7-6+
Formula Weight 142.0994
Chemical Name or Material trans-2-Octenoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.