accessibility menu, dialog, popup

UserName

CAS RN 18721-61-4

IUPAC Name: 3-(ethylsulfanyl)propan-1-ol
Synonyms: 3-(ethylsulfanyl)propan-1-ol
Molecular Weight (g/mol): 120.21
Molecular Formula: C5H12OS
InChi Key: KRUJXSIEMOWXOF-UHFFFAOYSA-N
SMILES: CCSCCCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results