accessibility menu, dialog, popup

UserName

CAS RN 187618-60-6

H3CCH3SOONHNOHOHNOONH2CH3OCH3H3C(E/Z)

CAS RN 187618-60-6

IUPAC Name: 5-{[amino(2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-sulfonamido)methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 5-{[amino(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-sulfonamido)methylidene]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 648.78
Molecular Formula: C34H40N4O7S
InChi Key: HNICLNKVURBTKV-UHFFFAOYNA-N
SMILES: CC1=C2CC(C)(C)OC2=C(C)C(C)=C1S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results