CAS RN 187978-79-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-(3-Methylphenyl)imidazolidine-2,4-dione, TRC
CAS: 187978-79-6 Molecular Formula: C10H10N2O2 Molecular Weight (g/mol): 190.2 Synonym: 5-(3-Methylphenyl)-2,4-imidazolidinedione IUPAC Name: 5-(3-methylphenyl)imidazolidine-2,4-dione SMILES: Cc1cccc(c1)C2NC(=O)NC2=O