missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Methylphenyl)imidazolidine-2,4-dione, TRC

Product Code. 30477917
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477917 500 mg 500mg
30474561 50 mg 50mg
30476876 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477917 Supplier Toronto Research Chemicals Supplier No. TRCM259756500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 187978-79-6
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.2
Synonym 5-(3-Methylphenyl)-2,4-imidazolidinedione
IUPAC Name 5-(3-methylphenyl)imidazolidine-2,4-dione
SMILES Cc1cccc(c1)C2NC(=O)NC2=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H10N2O2/c1-6-3-2-4-7(5-6)8-9(13)12-10(14)11-8/h2-5,8H,1H3,(H2,11,12,13,14)
Formula Weight 190.0742
Chemical Name or Material 5-(3-Methylphenyl)imidazolidine-2,4-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.