accessibility menu, dialog, popup

UserName

HBrN

CAS RN 18820-82-1

IUPAC Name: hydrogen pyridine bromide
Synonyms: pyridine, hydrogen bromide
Molecular Weight (g/mol): 160.01
Molecular Formula: C5H6BrN
InChi Key: BBFCIBZLAVOLCF-UHFFFAOYSA-N
SMILES: [H+].[Br-].C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (5)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (3)
  • (5)
  • (9)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (4)