missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrobromide, 97%, BLD Pharm™

Product Code. 30801730
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30801730 1 kg 1kg
30800484 10 g 10g
30789390 100 g 100g
30792060 25 g 25g
30790364 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30801730 Supplier BLD Pharm Supplier No. BD1497021000g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18820-82-1
Molecular Formula C5H6BrN
MDL Number MFCD00012801
Quantity 1 kg
SMILES [H]Br.C1=NC=CC=C1
Molecular Weight (g/mol) 160.01
Percent Purity 97%
Chemical Name or Material Pyridine hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.