accessibility menu, dialog, popup

UserName

CAS RN 1883-09-6

HHClClH2NNH2OHO

CAS RN 1883-09-6

IUPAC Name: dihydrogen 2,4-diaminobutanoic acid dichloride
Synonyms: dihydrogen 2,4-diaminobutanoic acid dichloride
Molecular Weight (g/mol): 191.05
Molecular Formula: C4H12Cl2N2O2
InChi Key: CKAAWCHIBBNLOJ-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCC(N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)