missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-2,4-Diaminobutyric Acid, Dihydrochloride, TRC
Specifications
Specifications
| CAS | 1883-09-6 |
| Molecular Formula | C4 H10 N2 O2 . 2 Cl H |
| Quantity | 1 g |
| Synonym | Butanoic acid, 2,4-diamino-, hydrochloride (1:2), (2S)- (ACI), Butanoic acid, 2,4-diamino-, dihydrochloride, (2S)- (9CI), Butanoic acid, 2,4-diamino-, dihydrochloride, (S)- (ZCI), Butyric acid, 2,4-diamino-, dihydrochloride, L- (8CI), (2S)-2,4-Diaminobutanoic acid dihydrochloride, L-2,4-Diaminobutanoic acid dihydrochloride, L-2,4-Diaminobutyric acid dihydrochloride, L-Dab-OH dihydrochloride |
| InChI Formula | InChI=1S/C4H10N2O2.2ClH/c5-2-1-3(6)4(7)8;;/h3H,1-2,5-6H2,(H,7,8);2*1H/t3-;;/m0../s1 |
| SMILES | Cl.Cl.NCC[C@H](N)C(=O)O |
| IUPAC Name | (2S)-2,4-diaminobutanoic acid;dihydrochloride |
| Molecular Weight (g/mol) | 191.06 |
| Formula Weight | 190.0276 |
| Chemical Name or Material | L-2,4-Diaminobutyric Acid, Dihydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?