accessibility menu, dialog, popup

UserName

CAS RN 1883-32-5

OH

CAS RN 1883-32-5

IUPAC Name: 2,2-diphenylethan-1-ol
Synonyms: 2,2-diphenyl ethanol
Molecular Weight (g/mol): 198.27
Molecular Formula: C14H14O
InChi Key: NYLOEXLAXYHOHH-UHFFFAOYSA-N
SMILES: OCC(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results