accessibility menu, dialog, popup

UserName

H3CH3CCH3OON

CAS RN 188345-71-3

IUPAC Name: tert-butyl 2-azabicyclo[2.2.1]hept-5-ene-2-carboxylate
Synonyms: tert-butyl 2-azabicyclo[2.2.1]hept-5-ene-2-carboxylate
Molecular Weight (g/mol): 195.26
Molecular Formula: C11H17NO2
InChi Key: ZATXHTCUZAWODK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC2CC1C=C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results