missing translation for 'onlineSavingsMsg'
Learn More

2-Boc-2-azabicyclo[2.2.1]hept-5-ene, 98%

Product Code. 15450529
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450529 5 g 5g
15460529 250 mg 250mg
15440529 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450529 Supplier Thermo Scientific Alfa Aesar Supplier No. H63698.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 188345-71-3 | C11H17NO2 | 195.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188345-71-3
Molecular Formula C11H17NO2
Molecular Weight (g/mol) 195.262
MDL Number MFCD22416354
InChI Key ZATXHTCUZAWODK-UHFFFAOYSA-N
PubChem CID 15837921
IUPAC Name tert-butyl 5-azabicyclo[2.2.1]hept-2-ene-5-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2CC1C=C2

Specifications

Color Yellow
Quantity 5 g
Synonym 2-boc-2-azabicyclo 2.2.1 hept-5-ene, tert-butyl 2-aza-bicyclo 2.2.1 hept-5-ene-2-carboxylate, tert-butyl 2-azabicyclo 2.2.1 hept-5-ene-2-carboxylate, 2-tert-butoxycarbonyl-2-azabicyclo 2.2.1 hept-5-ene, tert-butyl2-azabicyclo 2.2.1 hept-5-ene-2-carboxylate, ksc497c1n, tert-butyl 5-azabicyclo 2.2.1 hept-2-ene-5-carboxylate, 2-aza-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid tert-butyl ester, 2-tert-butyloxycarbonyl-2-azabicyclo 2.2.1 hepta-5-ene, 2-azabicyclo 2.2.1 hept-5-ene-2-carboxylic acid tert-butyl ester
Formula Weight 195.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Boc-2-azabicyclo[2.2.1]hept-5-ene

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.