accessibility menu, dialog, popup

UserName

CAS RN 18865-74-2

CH3H3CCH3OCH3H3CCH3OHCH3H3CCH3OCH3H3CCH3OHCH3H3CCH3OCH3H3CCH3OHCH3H3CCH3OCH3H3CCH3OHZr(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 18865-74-2

IUPAC Name: tetrakis(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) zirconium
Synonyms: tetrakis(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) zirconium
Molecular Weight (g/mol): 828.34
Molecular Formula: C44H80O8Zr
InChi Key: ANBXKEDJQLYCLX-UHFFFAOYSA-N
SMILES: [Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results