missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11349697
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349697 25 g 25g
11329697 1 g 1g
11339697 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349697 Supplier Thermo Scientific Alfa Aesar Supplier No. 042028.14
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

MOCVD of zirconium

MOCVD of zirconium

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18865-74-2
Molecular Formula C44H80O8Zr
Molecular Weight (g/mol) 828.34
MDL Number MFCD00145380,MFCD00145380
InChI Key ANBXKEDJQLYCLX-UHFFFAOYSA-N
Synonym tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one
PubChem CID 50919870
IUPAC Name 5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium
SMILES [Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C

Specifications

Melting Point 318°C to 320°C
Boiling Point 180°C (0.1 mmHg)(sublimation)
Quantity 25 g
Formula Weight 824.3
Percent Purity 99.99%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.