accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OMgOHOOOHOO

CAS RN 18917-95-8

IUPAC Name: magnesium(2+) bis(2-hydroxybenzoate) tetrahydrate
Synonyms: magnesium(2+) tetrahydrate disalicylate
Molecular Weight (g/mol): 370.59
Molecular Formula: C14H18MgO10
InChi Key: NBQBEWAYWAMLJJ-UHFFFAOYSA-L
SMILES: O.O.O.O.[Mg++].OC1=CC=CC=C1C([O-])=O.OC1=CC=CC=C1C([O-])=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results