CAS RN 189628-37-3
IUPAC Name:
5-bromo-2-chlorobenzaldehyde
Synonyms:
5-bromo-2-chlorobenzaldehyde
Molecular Weight (g/mol):
219.46
Molecular Formula:
C7H4BrClO
InChi Key:
DPKKRQAEYWOISP-UHFFFAOYSA-N
SMILES:
ClC1=C(C=O)C=C(Br)C=C1
1
–
4
of
4
results
1
5-Bromo-2-chlorobenzaldehyde, 98%, Thermo Scientific Chemicals
CAS: 189628-37-3 Molecular Formula: C7H4BrClO Molecular Weight (g/mol): 219.46 MDL Number: MFCD08445659 InChI Key: DPKKRQAEYWOISP-UHFFFAOYSA-N PubChem CID: 10608925 IUPAC Name: 5-bromo-2-chlorobenzaldehyde SMILES: ClC1=C(C=O)C=C(Br)C=C1
2
5-Bromo-2-chlorobenzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Bromo-2-chlorobenzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
5-Bromo-2-chlorobenzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.