CAS RN 190728-25-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(6,7-Dimethoxy-quinolin-4-yloxy)-phenylamine, TRC
CAS: 190728-25-7 Molecular Formula: C17H16N2O3 Molecular Weight (g/mol): 296.32 Synonym: 4-[(6,7-Dimethoxy-4-quinolinyl)oxy]benzenamine,4-[(6,7-Dimethoxyquinolin-4-yl)oxy]aniline,6,7-Dimethoxy-4-(4-aminophenoxy)quinoline;[4-[(6,7-Dimethoxyquinolin-4-yl)oxy]phenyl]amine; SMILES: COc1cc2nccc(Oc3ccc(N)cc3)c2cc1OC