missing translation for 'onlineSavingsMsg'
Learn More

4-(6,7-Dimethoxy-quinolin-4-yloxy)-phenylamine, TRC

Product Code. 30011152
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011152 5 g 5g
30007552 10 g 10g
30010258 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011152 Supplier Toronto Research Chemicals Supplier No. TRCD4630455G
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190728-25-7
Molecular Formula C17H16N2O3
Molecular Weight (g/mol) 296.32
Synonym 4-[(6,7-Dimethoxy-4-quinolinyl)oxy]benzenamine, 4-[(6,7-Dimethoxyquinolin-4-yl)oxy]aniline, 6,7-Dimethoxy-4-(4-aminophenoxy)quinoline;[4-[(6,7-Dimethoxyquinolin-4-yl)oxy]phenyl]amine;
SMILES COc1cc2nccc(Oc3ccc(N)cc3)c2cc1OC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C17H16N2O3/c1-20-16-9-13-14(10-17(16)21-2)19-8-7-15(13)22-12-5-3-11(18)4-6-12/h3-10H,18H2,1-2H3
Chemical Name or Material 4-(6,7-Dimethoxy-quinolin-4-yloxy)-phenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.