accessibility menu, dialog, popup

UserName

CH3SnH3CCH3NNH3CFFF

CAS RN 191606-78-7

IUPAC Name: 1-methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole
Synonyms: 1-methyl-5-(tributylstannyl)-3-(trifluoromethyl)pyrazole
Molecular Weight (g/mol): 439.15
Molecular Formula: C17H31F3N2Sn
InChi Key: KZKSCFOBDCKLCL-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=CC(=NN1C)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results