missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10423223
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10423223 250 mg Amber Glass Bottle 250mg
10399042 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10423223 Supplier Thermo Scientific Maybridge Supplier No. CC29541CB
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 191606-78-7
Molecular Formula C17H31F3N2Sn
Molecular Weight (g/mol) 439.154
MDL Number MFCD06797466
InChI Key KZKSCFOBDCKLCL-UHFFFAOYSA-N
Synonym 1-methyl-5-tributylstannyl-3-trifluoromethyl-1h-pyrazole, 1-methyl-3-trifluoromethyl-5-tributylstannylpyrazole, 1-methyl-5-tributylstannyl-3-trifluoromethyl pyrazole, tributyl-2-methyl-5-trifluoromethyl pyrazol-3-yl stannane, 1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl tributylstannane, 1h-pyrazole,1-methyl-5-tributylstannyl-3-trifluoromethyl, tributyl-2-methyl-5-trifluoromethyl-3-pyrazolyl stannane, 1-methyl-5 tributylstannyl 3-trifluoromethyl-1h-pyrazole, 1-methyl-5-tributylstannanyl-3-trifluoromethyl-1h-pyrazole
PubChem CID 12056710
IUPAC Name tributyl-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC(=NN1C)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 439.15
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-(tributylstannyl)-3-(trifluoromethyl)-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.