accessibility menu, dialog, popup

UserName

CAS RN 19241-39-5

CH3NCSBr

CAS RN 19241-39-5

IUPAC Name: 2-bromo-1-isothiocyanato-4-methylbenzene
Synonyms: 2-bromo-1-isothiocyanato-4-methylbenzene
Molecular Weight (g/mol): 228.11
Molecular Formula: C8H6BrNS
InChi Key: DQXSCOWMTPPFCJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(N=C=S)C(Br)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results