accessibility menu, dialog, popup

UserName

CH3OOBr

CAS RN 1927-95-3

IUPAC Name: 4-bromophenyl acetate
Synonyms: 4-bromophenyl acetate
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: XEXHCQJRVMVJMY-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results