accessibility menu, dialog, popup

UserName

CAS RN 193290-19-6

IUPAC Name: methyl 2-(4-bromo-2-fluorophenyl)acetate
Synonyms: methyl 2-(4-bromo-2-fluorophenyl)acetate
Molecular Weight (g/mol): 247.06
Molecular Formula: C9H8BrFO2
InChi Key: QBGQCRUYARHQGO-UHFFFAOYSA-N
SMILES: COC(=O)CC1=C(F)C=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results