accessibility menu, dialog, popup

UserName

HOBOHFF

CAS RN 193353-34-3

IUPAC Name: (2,5-difluorophenyl)boronic acid
Synonyms: 2,5-difluorophenylboronic acid
Molecular Weight (g/mol): 157.91
Molecular Formula: C6H5BF2O2
InChi Key: KTOJGSDLJNUAEP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=CC=C1F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results