accessibility menu, dialog, popup

UserName

CAS RN 193887-44-4

CH3H3C(S)HOONHOO

CAS RN 193887-44-4

IUPAC Name: (3S)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-methylhexanoic acid
Synonyms: (3S)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-methylhexanoic acid
Molecular Weight (g/mol): 367.45
Molecular Formula: C22H25NO4
InChi Key: YLVSABQQLLRFIJ-HNNXBMFYSA-N
SMILES: CC(C)C[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results