accessibility menu, dialog, popup

UserName

CAS RN 1940-57-4

IUPAC Name: 9-bromo-9H-fluorene
Synonyms: fluorene, 9-bromo-
Molecular Weight (g/mol): 245.12
Molecular Formula: C13H9Br
InChi Key: AHCDKANCCBEQJJ-UHFFFAOYSA-N
SMILES: BrC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)